Learn More
Applied Biosystems™ POP-7™ Polymer, for 3500/SeqStudio™ Flex
Catalog No. 4393708
Encompass
Change view
Click to view available options
Quantity:
384 Samples
96 Samples
960 Samples
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4393708 384 Samples
A26073 96 Samples
4393714 960 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4393708 Supplier Applied Biosystems™ Supplier No. 4393708
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
The pouch contains 384 samples of the polymer. Store the pouch at 2 to 8°C until ready to use. Refer to the expiration date on the label. Note that expired pouches cannot be used on the instrument.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages. The POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages, POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis. POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis.
• Easy to use: pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
• Include radio frequency identification (RFID) label: easy to track important information such as part⁄lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation: formulated for sequencing and fragment analysis applications
• Replenishable: allow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID⁄Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.
Order Info
Each
Specifications
Quantity 384 Samples
For Use With (Application) Long-Read Sequencing, Short Read Sequencing, Fragment Analysis (Denaturing)
Product Line POP-7
What is the difference between the compositions of POP-7 in POP-7 Polymer for Protein Quality Analysis (Cat. Nos. A30936, A31122) and POP-7 Polymer, for 3500/SeqStudio Flex (Cat. Nos. 4393708, A26703, 4393714)?
There is no difference between the compositions of POP-7 in POP-7 Polymer for Protein Quality Analysis (Cat. Nos. A30936, A31122) and POP-7 Polymer, for 3500/SeqStudio Flex (Cat. Nos. 4393708, A26703, 4393714) as they come from the same manufacturing bulk/formulation.
What is the white residue around the Applied Biosystems 3500/3500xL Genetic Analyzer array knob?
When placing the array on the instrument, the last step in the process is to loosen the array knob and the instrument forces any air bubbles around the array tip through the threading and the middle of the array knob. Some polymer gets pushed through as well and when it dries up, it appears as a white, flaky residue. Use a damp, lint-free cloth to remove it.
I seem to be getting bubbles from the polymer pouch on my Applied Biosystems 3500/3500xL Genetic Analyzer but the polymer level is normal. What is causing it?
On the Applied Biosystems 3500 Series Genetic Analyzer, the polymer pouch may not be sitting properly or the check valve may be stuck in the open position. Carefully detach the polymer pouch by pulling down the handle next to it, and then pull the handle up while guiding the pouch into place and secure it by pushing up on the plastic head of the pouch. If this fails, perform a wash on the system using the conditioning pouch. If reseating the tube/pouch and cleanup do not fix the problem, a service call may be required.
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
I have an Applied Biosystems 3500/3500xL Genetic Analyzer. When do my consumables expire?
If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.
My Applied Biosystems 3500/3500xL Genetic Analyzer consumables have expired, but how long past expiration can I really use them?
Thermo Fisher Scientific can only provide warranty on performance on non-expired reagents. Using reagents past their expiration date is a risk and may compromise your data integrity.
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
How long can I keep the buffer on the Applied Biosystems 3500/3500xL Genetic Analyzer?
For the Applied Biosystems 3500 Series Genetic Analyzer, the buffer will keep for 14 days if room temperature is <25 degrees C and for 7 days if >25 degrees C.