Learn More
Brady™ Polystyrene Sign: CAUTION - HOT SURFACE. DO NOT TOUCH.
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Supplier:  Brady™ 143822
 Afficher plus de versions de ce produit
Numéro de catalogue. 19110715
Il en reste null
Description
Description
  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Spécifications
Identifiants chimiques
Polystyrene
Rectangular
CAUTION - HOT SURFACE. DO NOT TOUCH.
Black on White
Spécifications
Heat Picto
Rectangular
CAUTION - HOT SURFACE. DO NOT TOUCH.
Polystyrene
14 x 10 in.
Black on White
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS