missing translation for 'onlineSavingsMsg'
Learn More

Polystyrène, MilliporeSigma™ Supelco™

Numéro de catalogue. 111004507
Modifier l'affichage
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111004507 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111004507 Fournisseur Merck Emd Millipore Code fournisseur 18243525G

Spécifications

Density 1.05 g/mL (at 25°C)
Physical Form Pellets
Linear Formula [CH2 CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Type Analytical Standard
Quantity 25 g
Grade Norme analytique