missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polystyrene for GPC 150,000, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Physical Form | Neat |
| Linear Formula | [CH2 CH(C6H5)]n |
| MDL Number | MFCD00084450 |
| Synonym | PS |
| Type | Analytical Standard |
| Quantity | 1 g |
| Grade | GPC |