missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polystyrene Containers

Ideal for storing dry materials, powders and tablets.

 Afficher plus de versions de ce produit

Numéro de catalogue. 033383D


Il en reste null

Description

Description

Ideal for storing dry materials, powders and tablets.

  • Made of polystyrene material
  • With snap-on caps

Spécifications

Identifiants chimiques

Polystyrene
Cylindrical
70°C
Snap Cap
Storing dry materials, powders, tablets
158°F
Polyethylene
Storage Vial with Cap

Spécifications

Container w Snap Cap 10 Dr
27 mm
Polystyrene
Single-use
Storing dry materials, powders, tablets
3.23 in.
Cylindrical
70°C
37 mL
Snap Cap
Not Autoclavable
Storage Vial with Cap
1.063 in.
Transparent
Non-sterile
37 mL
No
Snap Cap
158°F
10 dr.
Polyethylene
144/Cs.
82 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions