missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80

Catalog No. AAL13315AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13315AE 100 mL
AAL13315AP 500 mL
AAL133150F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13315AE Supplier Thermo Scientific Chemicals Supplier No. L13315AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol

Used for solubilizing membrane proteins during isolation and purification.Polysorbate 80 is used as a surfactant in soaps, cosmetics, body washes, hair shampoos, hair loss treatments, lotions, creams, bath oils and in mouthwash in the health care industry. It serves as an emulsifier in foods such as ice cream and in the preparation of antiarrhythmic amiodarone. Further, it acts as a solubilizing agent and excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
Synonym Polyoxyethylene sorbitan monooleate
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Boiling Point >100°C
Density 1.064
Quantity 100 mL
Flash Point >149°C (300°F)
Refractive Index 1.475
Merck Index 14,7582
Solubility Information Miscible with water,ethanol,cottonseed oil,corn oil,ethyl acetate,methanol and toluene. Immiscible with mineral and vegetable oils.
Chemical Name or Material Polysorbate 80
RTECSNumber WG2932500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only