missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80, NF, Spectrum™ Chemical

PO138, 9005-65-6

$1,687.79 - $19,139.82

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18605394
View Documents Promotion Details
N/A 6 x 500 mL Amber Glass Bottle
Case of 6 Each for $2,718.96
Only null left
 
18605395
View Documents Promotion Details
N/A 4 L Amber Glass Bottle
Each for $1,687.79
Only null left
 
18605396
View Documents Promotion Details
N/A 4 x 4 L Amber Glass Bottle
Case of 4 Each for $5,861.11
Only null left
 
18605397
View Documents Promotion Details
N/A 20 L Amber Glass Bottle
Each for $4,257.00
Only null left
 
18605398
View Documents Promotion Details
N/A 7 gal. Amber Glass Bottle
Each for $8,033.07
Only null left
 
18605399
View Documents Promotion Details
N/A 200 L Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Polysorbate 80, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

9005-65-6
604.82
HDTIFOGXOGLRCB-KTKRTIGZNA-N
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MFCD00082107
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate

Specifications

9005-65-6
1
65-80
45-55
300-500 cSt
MFCD00082107
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
604.82
Polysorbate 80
2
Neutral
0.0025
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
0.03
HDTIFOGXOGLRCB-KTKRTIGZNA-N
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
NF
Videos
SDS
Documents

Documents