missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 80, NF, Spectrum™ Chemical
PO138, 9005-65-6
Supplier: Spectrum Chemical Mfg Cor PO1384LTGLCS4
| Quantity | 4 x 4 L |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Polysorbate 80, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 9005-65-6 | |
| 604.82 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| MFCD00082107 | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
Specifications
| Polysorbate 80 | |
| 2 | |
| Neutral | |
| 65-80 | |
| Amber Glass Bottle | |
| (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
| 0.03 | |
| HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
| 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
| NF |
| 9005-65-6 | |
| 1 | |
| 4 x 4 L | |
| 0.0025 | |
| 45-55 | |
| 300-500 cSt | |
| MFCD00082107 | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 604.82 |