missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80, NF, Spectrum™ Chemical

Catalog No. 18605394
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
20 L
7 gal.
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605394 6 x 500 mL Amber Glass Bottle
18605395 4 L Amber Glass Bottle
18605396 4 x 4 L Amber Glass Bottle
18605397 20 L Amber Glass Bottle
18605398 7 gal. Amber Glass Bottle
18605399 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18605394 Supplier Spectrum Chemical Mfg Cor Supplier No. PO138500MLGLCS6

PO138, 9005-65-6

Spectrum™ Chemical Polysorbate 80, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Acid Value 2
CAS Max % 1
pH Neutral
Quantity 6 x 500 mL
Hydroxyl Value 65-80
Ignition Residue 0.0025
Packaging Amber Glass Bottle
Saponification Value 45-55
Viscosity 300-500 cSt
Water 0.03
Grade NF
Chemical Name or Material Polysorbate 80
Show More Show Less