missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 60
CAS: 9005-67-8 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | 522.68 g/mol
Supplier: Thermo Scientific Chemicals 278620010
| Quantity | 1 L |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 9005-67-8 | |
| 522.68 | |
| DEKFZWMENCCOFU-UHFFFAOYNA-N | |
| CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
| MFCD01780110 | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
Spécifications
| Polysorbate 60 | |
| 2mg KOH/g max. | |
| Paste at 25°C | |
| 1 L | |
| 81 to 96mg KOH/g | |
| Glass bottle | |
| 1.08 | |
| 75-175 Pa.s (50°C) | |
| MFCD01780110 | |
| DEKFZWMENCCOFU-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| Yellow | |
| 9005-67-8 | |
| 1.0800g/mL | |
| >150°C | |
| Authentic | |
| 45 to 55mg KOH/g | |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
| 3% max. | |
| Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, xylene, insoluble in propylene glycol and mineral oil | |
| CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 |