missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 40, NF, Spectrum™ Chemical

Catalog No. 18605384
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605384 25 mL Amber Glass Bottle
18605380 500 mL Amber Glass Bottle
18605381 4 L Amber Glass Bottle
18605382 20 L Amber Glass Bottle
18605383 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605384 Supplier Spectrum Chemical Mfg Cor Supplier No. PO13425ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PO134, 9005-66-7

Spectrum™ Chemical Polysorbate 40, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-66-7
Molecular Formula (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165345
InChI Key DVRFSZFBSOKHQW-UHFFFAOYNA-N
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 25 mL
Hydroxyl Value 89-105
Ignition Residue 0.0025
Packaging Amber Glass Bottle
Saponification Value 41-52
Water 0.03
Grade NF
Chemical Name or Material Polysorbate 40