missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 20
CAS: 9005-64-5 | C26H50O10
Supplier: Thermo Scientific Chemicals 233360051
| Quantity | 5 L |
|---|---|
| Packaging Type | Plastic Bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9005-64-5 | |
| C26H50O10 | |
| Polyoxyethylene(20)sorbitan monolaurate |
Specifications
| Polysorbate 20 | |
| Colorless to Yellow | |
| C26H50O10 | |
| 5 L | |
| Polyoxyethylene(20)sorbitan monolaurate |
| 9005-64-5 | |
| 1.1000 g/mL | |
| Plastic Bottle | |
| Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol | |
| Liquid |