missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20

Catalog No. AC233360050
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
5 L
25 L
Packaging Type:
Glass Bottle
Plastic Bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
AC233360050 5 mL Glass Bottle
AC233362500 250 mL Glass Bottle
AC233360010 1 L Glass Bottle
AC233360051 5 L Plastic Bottle
AC233360250 25 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC233360050 Supplier Thermo Scientific Chemicals Supplier No. 233360050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 9005-64-5 | C26H50O10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula C26H50O10
Synonym Polyoxyethylene(20)sorbitan monolaurate

Specifications

Color Colorless to Yellow
Density 1.1000 g/mL
Quantity 5 mL
Format Liquid
Packaging Type Glass Bottle
Solubility Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol
Form Liquid
Chemical Name or Material Polysorbate 20

RUO – Research Use Only