missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 20, Multi-Compendial, N.F., J.T. Baker™
$2,267.00 - $177,893.89
Chemical Identifiers
| CAS | 9005-64-5 |
|---|---|
| Molecular Formula | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 |
| Molecular Weight (g/mol) | 522.68 |
| MDL Number | MFCD00165986 |
| InChI Key | HMFKFHLTUCJZJO-UHFFFAOYNA-N |
| PubChem CID | 443314 |
| SMILES | CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
02003671
|
Avantor J.T.Baker
411602 |
20 mL | Narrow Mouth Amber Glass Bottle |
Each for $2,267.00
|
|
||||
|
02003669
|
Avantor J.T.Baker
411604 |
200 mL | Narrow Mouth Amber Glass Bottle |
Each for $3,046.78
|
|
||||
|
02003665
|
Avantor J.T.Baker
411606 |
1 L | Narrow Mouth Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
02003672
|
Avantor J.T.Baker
411603 |
4 L | Narrow Mouth Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
02003664
|
Avantor J.T.Baker
411607 |
19 L | Steel Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 9005-64-5 | |
| 522.68 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| MFCD00165986 | |
| 443314 |
Specifications
| 9005-64-5 | |
| 1L = 1.11kg | |
| MFCD00165986 | |
| CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 | |
| Multi-Compendial/NF |
| 148.00°C | |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
| HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
| 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
| 443314 | |
| Polysorbate 20 |