missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Multi-Compendial, N.F., J.T. Baker™

Catalog No. 02003671
Change view
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Steel Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003671 20 mL Narrow Mouth Amber Glass Bottle
02003669 200 mL Narrow Mouth Amber Glass Bottle
02003665 1 L Narrow Mouth Amber Glass Bottle
02003672 4 L Narrow Mouth Amber Glass Bottle
02003664 19 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02003671 Supplier Avantor Supplier No. 411602

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 20 mL
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.11kg
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 20