missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Polypropylene Petri Dishes with Covers

Eisco™ Polypropylene Petri Dishes with Covers are reusable and can be repeatedly autoclaved.

Supplier:  Eisco™ CH0372B

 View more versions of this product

Catalog No. S40603


Only null left

Description

Description

  • Made from high quality polypropylene (PP)
  • Includes dish and cover
  • Reusable
  • Autoclavable at 121°C

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Yes
Concave
Reusable
Polypropylene
Non-sterile
Petri Dish
Culturing
12.7 mm
2.95 in.
Autoclavable
Clear
Not Gridded
Round
Non-treated
1
1
0.5 in.
75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions