missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Polypropylene Petri Dishes with Covers

Eisco™ Polypropylene Petri Dishes with Covers are reusable and can be repeatedly autoclaved.

Supplier:  Eisco™ CH0372APK12

 View more versions of this product

Catalog No. S29421


Only null left

Description

Description

  • Made from high quality polypropylene (PP)
  • Includes dish and cover
  • Reusable
  • Autoclavable at 121°C

Warranty

1 year limited manufacturer warranty

Specifications

Chemical Identifiers

Yes
Clear
Not Gridded
Round
Non-treated
1
Autoclavable
Reusable
Polypropylene
Non-sterile
Petri Dish
Culturing

Specifications

Yes
Flat
Reusable
Polypropylene
Non-sterile
Petri Dish
Culturing
1.27 cm
2 in.
Autoclavable
Clear
Not Gridded
Round
Non-treated
1
12/Pk.
0.5 in.
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions