missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Polypropylene, Carboy with Spigot
GREENER_CHOICE

Catalog No. 19830440
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19830440 10 L
19830439 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19830440 Supplier Justrite™ Supplier No. JMC12939

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Constructed from autoclavable USP Class VI, FDA grade materials

Specifications

Accuracy ±5%
Autoclavable Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 330 x 163 x 495 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Height (English) 19.5 in.
Includes Spigot
Length (English) 13 in.
Material Polypropylene
Product Type Carboy with Spigot
Shape Rectangular
Width (English) 6.4 in.
Show More Show Less