missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Polypropylene, Carboy with Spigot
GREENER_CHOICE

Numéro de catalogue. 19830439
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (Metric)
19830439 5 L
19830440 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 19830439 Fournisseur Justrite™ Code fournisseur JMC12938
Il en reste null

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Constructed from autoclavable USP Class VI, FDA grade materials

Spécifications

Accuracy ±5%
Autoclavable Autoclavable
Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 605 x 132 x 414 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Height (English) 16.3 in.
Includes Spigot
Length (English) 12 in.
Material Polypropylene
Product Type Carboy with Spigot
Shape Rectangular
Width (English) 5.2 in.
Afficher plus de résultats Afficher moins de résultats