missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Polypropylene, Carboy with 83 mm Cap and Spigot
GREENER_CHOICE

Catalog No. 19830441
Change view
Click to view available options
Capacity (English):
5.3 gal.
Capacity (Metric):
20 L
Closure Size:
83 mm
Dimensions (L x W x H):
387 x 206 x 615 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric) Closure Size Dimensions (L x W x H)
19830441 5.3 gal. 20 L 83 mm 387 x 206 x 615 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19830441 Supplier Justrite™ Supplier No. JMC12940

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.; Autoclavable; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Constructed from autoclavable USP Class VI, FDA grade materials

Specifications

Accuracy ±5%
Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 387 x 206 x 615 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.
Height (English) 24.2 in.
Includes Spigot
Length (English) 15.25 in.
Material Polypropylene
Product Type Carboy with Spigot
Shape Rectangular
Width (English) 8.1 in.
Show More Show Less