missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Polypropylene, Carboy with Sanitary Neck

Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
Specifications
| Accuracy | ±5% |
| Autoclavable | Autoclavable |
| Capacity (English) | 10.6 gal. |
| Capacity (Metric) | 40 L |
| Color | White (Translucent) |
| Dimensions (L x W x H) | 434 x 249 x 737 mm |
| For Use With (Application) | For storing and dispensing solvents, buffers, reagents and powders. |
| Height (English) | 29 in. |
| Length (English) | 17.1 in. |
| Material | Polypropylene |
| Show More |