Learn More
Tradewinds Direct Inc 18x150mm Polyproylene Test Tubes. Pack of 250
Catalog No. 501173407
Encompass
Disposable 18x150mm polypropylene test tube. Test tubes have a maximum capacity of 25mL. 250/Pack
Catalog No. 501173407 Supplier Tradewinds Direct Inc Supplier No. DPTT18X150
Disposable 18x150mm polypropylene test tube. Test tubes have a maximum capacity of 25mL. 250/Pack