missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Polypeptone™ Peptone

Pancreatic digest of casein and peptic digest of animal tissue combined in equal parts

$226.04 - $3315.00

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Various bacteria, fungi, and mammalian cells
Product Line Polypeptone
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
297108
View Documents
Gibco™
297108
10 kg
Each for $3,315.00
Only null left
 
B11910
View Documents
Gibco™
211910
454 g
Each for $226.04
Only null left
 
Description

Description

A mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digest of animal tissue.
Specifications

Specifications

Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Dehydrated Culture Media
Polypeptone
Powder
Various bacteria, fungi, and mammalian cells
Animal Origin
Videos
SDS
Documents

Documents