missing translation for 'onlineSavingsMsg'
Learn More

Polynuclear Aromatic Hydrocarbons Mix, Certified Reference Material, 2,000 μg/mL, MilliporeSigma™ Supelco™

Catalog No. 111016452
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016452 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016452 Supplier Merck Emd Millipore Supplier No. CRM48905

2,000 μg/mL Each Component in Methylene Chloride: Benzene (1:1)

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 2,000 μg/mL each component in methylene chloride: benzene (1:1)
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Acenaphthene; Acenaphthylene; Anthracene; Benz[a]anthracene; Benzo[b]fluoranthene; Benzo[k]fluoranthene; Benzo[ghi]perylene; Benzo[a]pyrene; Chrysene; Dibenz[a,h]anthracene; Fluoranthene; Fluorene;Indeno[1,2,3-cd]pyrene; Naphthalene; Phenanthrene; Pyrene

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC