missing translation for 'onlineSavingsMsg'
Learn More

Polyethyleneimine, 50% Aqueous Solution, MP Biomedicals™

Numéro de catalogue. ICN19544480
Modifier l'affichage
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ICN19544480 100 mL
ICN19544490 500 mL
ICN19544491 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. ICN19544480 Fournisseur MP Biomedicals Inc Code fournisseur 0219544480
Il en reste null

Identifiants chimiques

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*

Spécifications

Physical Form Viscous Liquid
Quantity 100 mL
Formula Weight 50,000 to 100,000
Chemical Name or Material Polyethyleneimine