missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol, MP Biomedicals™
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Density | 1.1 to 1.2g/mL |
| Quantity | 1 kg |
| Formula Weight | 8000 |
| Chemical Name or Material | Polyethylene glycol |