missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monostearate (n=approx. 25) (palmitate and stearate mixture), TCI America™

Catalog No. P072025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P072025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P072025G Supplier TCI America Supplier No. P072025G

Chemical Identifiers

CAS 9004-99-3
Molecular Formula C20H40O3
Molecular Weight (g/mol) 328.537
MDL Number MFCD00148007
InChI Key RFVNOJDQRGSOEL-UHFFFAOYSA-N
PubChem CID 24762
IUPAC Name 2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCO

Specifications

Color White
Quantity 25 g
Physical Form Crystalline Lumps
Chemical Name or Material Polyethylene Glycol Monostearate (n=approx. 25) (palmitate and stearate mixture)
EINECSNumber (7)-0088
RTECSNumber TQ5400000
TSCA Yes