missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monomethyl Ether 400, TCI America™

Catalog No. P2183100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2183100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2183100G Supplier TCI America Supplier No. P2183100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9004-74-4
Molecular Formula (C2H4O)nCH4O
Molecular Weight (g/mol) 76.10
MDL Number MFCD00084416
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethan-1-ol
SMILES COCCO

Specifications

Color Yellow
Quantity 100 g
Physical Form Liquid
Chemical Name or Material Polyethylene Glycol Monomethyl Ether 400
EINECSNumber (7)-0097&(2)-0444
RTECSNumber TR1581750
TSCA Yes