missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monododecyl Ether (n=approx. 25) [for Biochemical Research], TCI America™

Catalog No. P17775G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17775G Supplier TCI America Supplier No. P17775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyethylene Glycol Monolauryl Ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color White
Quantity 5 g
Physical Form Crystals
Chemical Name or Material Polyethylene Glycol Monododecyl Ether (n=approx. 25) [for Biochemical Research]
EINECSNumber (7)-0097
RTECSNumber JR5990000
TSCA Yes