missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Monocetyl Ether (n=approx. 23) [for Biochemical Research], TCI America™

Catalog No. P17765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17765G Supplier TCI America Supplier No. P17765G
Only null left

Chemical Identifiers

CAS 9004-95-9
Molecular Formula C56H114O21
Molecular Weight (g/mol) 1123.51
MDL Number MFCD00080892
InChI Key NLMKTBGFQGKQEV-UHFFFAOYSA-N
Synonym cetomacrogol 1000, brij-58, polyoxyethylene 20 cetyl ether, polyethylene glycol hexadecyl ether, polyethylene glycol 20 hexadecyl ether, 2-2-2-2-2-2-2-2-2-2-2-2-2-2-2-2-2-2-2-2-hexadecoxyethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethoxy ethanol, cetomacrogolum 1000, unii-iv5hor6sht, unii-lf9x1pn3xj, unii-5kly4iog20
PubChem CID 2724259
IUPAC Name 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol
SMILES CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White
Physical Form Crystal-Lump, Wax at 20°C
Quantity 5 g
Chemical Name or Material Polyethylene Glycol Monocetyl Ether (n=approx. 23) [for Biochemical Research]
EINECSNumber (7)-0097
RTECSNumber TR1581470
TSCA Yes