missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol Mono-4-octylphenyl Ether (n=approx. 10) [for Biochemical Research], TCI America™

Catalog No. P17755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17755G Supplier TCI America Supplier No. P17755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym triton x-100, triton x, octoxynol, preceptin, triton x-45, triton x 45, octoxynol-3, octoxynol-9, hydrol sw, triton x 100
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless
Physical Form Clear Liquid at 20°C
Melting Point 6°C
Quantity 5 g
Chemical Name or Material Polyethylene Glycol Mono-4-octylphenyl Ether (n=approx. 10) [for Biochemical Research]
EINECSNumber (7)-0172
RTECSNumber MD0907700
TSCA Yes