missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol 400

Catalog No. AAB2199230
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2199230 250 g
AAB219920B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2199230 Supplier Thermo Scientific Chemicals Supplier No. B2199230

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used in a variety of pharmaceutical formulations. It finds application in Hewlett-Packard designjet printers as an ink solvent and a lubricant for the print heads. It is an important raw material for the manufacture of polyuretnae, PEG salts of lauric, oleic and stearic acids and latex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 400
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Melting Point 4°C to 8°C
Linear Formula H(OCH2CH2)nOH
Density 1.126
Merck Index 14,7568
Solubility Information Miscible with water,acetone,alcohols,benzene,glycerin,glycols and aromatic hydrocarbons. Slightly miscible with aliphatic hydrocarbons.
Quantity 250 g
Flash Point 240°C (464°F)
Odor Odorless
Refractive Index 1.466
Sensitivity Hygroscopic
Chemical Name or Material Polyethylene glycol 400
Show More Show Less
Hazard Category H227-H303-H315-H320-H333-H335
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
RTECSNumber TQ3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only