missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 200, J.T. Baker™
Description
- For Laboratory,Research,or Manufacturing Use
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 4 L |
| Formula Weight | 190 to 210 (Average) |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.15kg |
| Chemical Name or Material | Polyethylene Glycol 200 |