missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PEEK Sample Loops
Resists organic solvents and salt buffers commonly used in chromatography of biomolecules. Cytiva Life Sciences™ PEEK Sample Loops are an important accessory for chromatography applications.
Supplier: Cytiva 18114053
Description
- Wetted parts are resistant to organic solvents
- Wetted parts are resistant to salt buffers
- Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
- Polyetheretherketone (PEEK)
Chemical Identifiers
| Sample Loop | |
| PEEK | |
| 1.58 mm |
Specifications
| Sample Loop | |
| 1.58 mm | |
| 5 mL |
| PEEK | |
| 1 mm |