missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PEEK Sample Loops

Catalog No. p-8001217
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
Diameter (Metric) Inner:
0.75 mm
1 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Diameter (Metric) Inner
361041167 1 mL 0.75 mm
361041168 2 mL 0.75 mm
361041169 5 mL 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361041167 Supplier Cytiva Supplier No. 18111401

Resists organic solvents and salt buffers commonly used in chromatography of biomolecules. Cytiva Life Sciences™ PEEK Sample Loops are an important accessory for chromatography applications.

  • Wetted parts are resistant to organic solvents
  • Wetted parts are resistant to salt buffers
  • Except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran
  • Polyetheretherketone (PEEK)

Specifications

Type Sample Loop
Material PEEK
Connection Size (Metric) 1.58 mm
Diameter (Metric) Inner 0.75 mm
Volume (Metric) 1 mL