missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polyester Sign: ELECTRICAL HAZARD - AUTHORIZED PERSONNEL ONLY. SERIOUS SHOCK OR DEATH COULD RESULT.
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$13.62 - $35.04
Spécifications
| Material | Polyester |
|---|---|
| Shape | Rectangular |
| Displayed Text | ELECTRICAL HAZARD - AUTHORIZED PERSONNEL ONLY. SERIOUS SHOCK OR DEATH COULD RESULT. |
| Color | Yellow on White |
| Numéro de catalogue | Numéro du manufacturier. | Dimensions (L x W) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Dimensions (L x W) | Prix | Quantity & Availability | |||||
|
19109646
|
Brady™
144624 |
14 x 10 in. |
chaque for $35.04
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
19109645
|
Brady™
144623 |
10 x 7 in. |
chaque for $13.62
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Spécifications
| Polyester | |
| Rectangular | |
| ELECTRICAL HAZARD - AUTHORIZED PERSONNEL ONLY. SERIOUS SHOCK OR DEATH COULD RESULT. | |
| Yellow on White |