missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 6000

Catalog No. AA42501K2
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42501K2 1 L
AA42501AD 50 mL
AA42501AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42501K2 Supplier Thermo Scientific Chemicals Supplier No. 042501K2
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy is used as reagents and Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Viscous Liquid
Boiling Point >205°C
Melting Point <-40°C
Density 0.950 g/mL
Solubility Information Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water.
Quantity 1 L
Flash Point 315°C (599°F)
Odor Odorless
Refractive Index 1.4025
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 6,000
Show More Show Less
RTECSNumber TQ2690000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only