accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
FR
chevron-down
Choisir la langue
English
Français
Rechercher
Statut de la commande
Commande rapide
Soutien
Offres
chevron-down
Featured
Spotlight
Offres
Dollars Flex
New Lab Start-up
Voir toutes
Se connecter
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chemicals
Organic compounds
Organic Polymers
ICN217538
Structure Search
imprimer
Polyacrylonitrile, MP Biomedicals
Numéro de catalogue.
icn217538
Modifier l'affichage
Chip view icon
List view icon
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
Quantity
ICN217538
5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue.
ICN217538
Fournisseur
MP Biomedicals Inc
Code fournisseur
0521753805
Connectez-vous
ou
enregistrez-vous
pour vérifier votre prix et la disponibilité.
Quantité
Il en reste null
Ajouter au panier
Ajouter au panier
Description
Spécifications
missing translation for 'recommendations'
Videos
Documents
Certificats
Sécurité et manipulation
Safety Services
Promotions
Spécifications
Identifiants chimiques
CAS
25014-41-9
Molecular Formula
C3H3N
Molecular Weight (g/mol)
53.064
InChI Key
NLHHRLWOUZZQLW-UHFFFAOYSA-N
Synonym
acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox
PubChem CID
7855
ChEBI
CHEBI:28217
IUPAC Name
prop-2-enenitrile
SMILES
C=CC#N
Spécifications
Quantity
5 g
Chemical Name or Material
Polyacrylonitrile
missing translation for 'recommendations'
Documents