missing translation for 'onlineSavingsMsg'
Learn More

Poly(N-vinylcarbazole), TCI America™

Catalog No. P06565G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06565G Supplier TCI America Supplier No. P06565G

Chemical Identifiers

CAS 25067-59-8
Molecular Formula (C14H11N)n
Molecular Weight (g/mol) 193.25
MDL Number MFCD00134336
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
PubChem CID 15143
IUPAC Name 9-ethenyl-9H-carbazole
SMILES *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Poly(N-vinylcarbazole)
EINECSNumber (6)-1465
RTECSNumber FE6225480
TSCA Yes