missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(methyl methacrylate), sec. stand., approx. M.W. 90,000, fine granules
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Beads |
| Melting Point | >150.0°C |
| Density | 1.18 |
| Quantity | 25 g |
| Flash Point | 303°C |
| Infrared Spectrum | Authentic |
| Loss on Drying | 5% max. |
| Packaging | Glass bottle |
| Particle Size | Bead size (typically): 50 to 150 micron |
| Show More |
RUO – Research Use Only