Learn More
Poly(methyl acrylate), solution in toluene, average M.W. ca. 40000 (GPC), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 310695000
| Quantity | 500mg |
|---|---|
| Packaging | Glass bottle |
Description
Chemical Identifiers
| C17H16O3 | |
| MFCD00084441 | |
| 4-phenylmethoxyphenyl methyl prop-2-enoate | |
| (4-phenylmethoxyphenyl)methyl prop-2-enoate |
Specifications
| Poly(methyl acrylate) | |
| C17H16O3 | |
| MFCD00084441 | |
| 4-phenylmethoxyphenyl methyl prop-2-enoate | |
| PTMPEQAUGJJXJV-UHFFFAOYSA-N | |
| (4-phenylmethoxyphenyl)methyl prop-2-enoate | |
| 4206998 | |
| Authentic | |
| 1.22 |
| 9003-21-8 | |
| [-CH2CH(CO2CH3)-]n | |
| 1.2200g/mL | |
| 500mg | |
| C=CC(=O)OCC1=CC=C(C=C1)OCC2=CC=CC=C2 | |
| 268.312 | |
| 4°C | |
| Glass bottle |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapour.
Suspected of damaging the unborn child.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger