missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Poly-L-Ornithine Solution (0.01%)

Numéro de catalogue. A004C
Modifier l'affichage
Click to view available options
Quantité:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
A004C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A004C Fournisseur MilliporeSigma™ Code fournisseur A004C
Edge

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Used as coating to enhance cell attachment and adhesion to both plasticware and glass surfaces

  • Positively charged
  • Used in combination with laminin for neuronal and neural stem cell cultures
  • Varying molecular weight: Less viscous lower molecular weight (30,000 d), higher molecular weight (greater than 300,000 d)
  • Uses 78000 d, yielding solution viscosity for easy handling while providing sufficient binding sites for cell attachment

Spécifications

Concentration 0.01% w/v (0.1 mg/mL)
À utiliser avec (application) Cell Attachment and Proliferation, Neuroscience, Cell Culture
Quantité 100 mL
Type de produit Poly-L-Ornithine Solution
Contenu et stockage Store at -20°C for up to 4 months from date of receipt. Avoid multiple freeze thaw cycles to maintain product integrity.
Forme Liquid