missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene oxide), M.W. 250,000 to 400,000

Numéro de catalogue. p-186194
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC178600050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC178600050 Fournisseur Thermo Scientific Chemicals Code fournisseur 178600050
Il en reste null

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Spécifications

Color White
Physical Form Powder
Melting Point 65.0°C to 67.0°C
Linear Formula (-CH2CH2O-)n
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Chemical Name or Material Poly(ethylene oxide), Approximate M.W. 300.000
Afficher plus de résultats Afficher moins de résultats
RTECSNumber TQ4100000
TSCA TSCA

RUO – Research Use Only