missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Poly(acrylic acid)
Description
Poly (acrylic acid) (PAA) is hygroscopic, brittle and colorless in nature with Tg at nearly 106oC. At temperatures above 200 to 250oC, it loses water and becomes an insoluble crosslinked polymer anhydride. Solubility of dried PAA in water increases with rise in temperatures. Concentrated solutions of PAA in water is thixotropic in nature.Poly (acrylic acid) (PAA)/Poly methyl methacrylate copolymers are used for drug delivery.Other applications may include:to study solute diffusion in Polyvinyl alcohol/PAA copolymer hydrogelsynthesizing poly(N-isopropylacrylamide)-block-PAA copolymer which responds to both temperature and pH stimuliin preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs as thickening agent for adhesives.
Spécifications
Spécifications
| MDL Number | MFCD00084394 |
| Quantity | 250 g |
| Molecular Weight (g/mol) | 450000 |