missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific Chemicals

Catalog No. p-7059506
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4474577 0.1 g
AA4474502 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4474577 Supplier Thermo Scientific Chemicals Supplier No. 04474577

Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) and devices. It is also used as organic light emitting diode (OLED) materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104934-50-1
Molecular Formula (C10H14S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00217686
InChI Key DUFPJSOXRHVDOV-UHFFFAOYSA-N
Synonym poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
PubChem CID 22950249
IUPAC Name 3-hexyl-2,5-dimethylthiophene
SMILES CCCCCCC1=C(-*)SC(-*)=C1

Specifications

Physical Form Powder
Solubility Information Soluble in Chloroform and tetrahydrofuran
Quantity 0.1 g
Chemical Name or Material Poly (3-hexylthiophene-2,5-diyl), Regioregular, low metals
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only