missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(1-vinylpyrrolidone-co-Vinyl Acetate) (Copolymer, 3:7) (50% in Ethanol), TCI America™
Supplier: TCI America P038525G
Specifications
| Yellow | |
| Clear Liquid at 20°C | |
| C10H15NO3 | |
| 1170 | |
| FYUWIEKAVLOHSE-UHFFFAOYSA-N | |
| 1-ethenylpyrrolidin-2-one; ethenyl acetate | |
| 270885 |
| Poly(1-vinylpyrrolidone-co-Vinyl Acetate) (Copolymer, 3:7) (50% in Ethanol) | |
| 25086-89-9 | |
| MFCD00134018 | |
| 25 g | |
| CC(=O)OC=C.C=CN1CCCC1=O | |
| 197.23 |
Chemical Identifiers
| 25086-89-9 | |
| 197.23 | |
| FYUWIEKAVLOHSE-UHFFFAOYSA-N | |
| 1-ethenylpyrrolidin-2-one; ethenyl acetate |
| C10H15NO3 | |
| MFCD00134018 | |
| 270885 | |
| CC(=O)OC=C.C=CN1CCCC1=O |
Safety and Handling
EINECSNumber : (6)-0326
RTECSNumber : AH3539000
TSCA : Yes