missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 188

Catalog No. AAJ6608736
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6608736 500 g
AAJ66087A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6608736 Supplier Thermo Scientific Chemicals Supplier No. J6608736

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9003-11-6 | C5H10O2

Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Poloxamer is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

Molecular Formula C5H10O2
Synonym Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer

Specifications

CAS 9003-11-6
Color White
Content And Storage Ambient temperatures
Quantity 500 g
Solubility Miscible with water.
Form Solid
Chemical Name or Material Poloxamer 188

RUO – Research Use Only