missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 188, Surfactant, Spectrum™ Chemical

Catalog No. 18612404
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612404 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18612404 Supplier Spectrum Chemical Mfg Cor Supplier No. P116112KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9003-11-6

Specifications

CAS Max % 1
Cloud Point >100°C
pH 6.0 to 7.0
Quantity 12 kg
Packaging Fiber Drum
Water 0.0075
Grade Ungraded
Chemical Name or Material Poloxamer 188

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.