missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PNU 74654

β-catenin binder; inhibits Wnt signaling

$223.64 - $223.64

Specifications

Chemical Name or Material 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide
CAS 113906-27-7
Assay Percent Range >99%
Molecular Formula C19H16N2O3
Formula Weight Observed MW: 320.34
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
35-345
View Documents
Tocris Bioscience™
35345
5 mg
Each for $223.64
Pack of 6 Each for $1,341.84
Only null left
 
Description

Description

Binds to β-catenin (KD = 450 nM). Inhibits the interaction between β-catenin and T cell factor 4 (Tcf4) and disrupts the Wnt signaling pathway.
Specifications

Specifications

2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide
>99%
Observed MW: 320.34
113906-27-7
C19H16N2O3
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'