accessibility menu, dialog, popup

UserName

CAS RN 113906-27-7

CH3ONNHOO(E/Z)

CAS RN 113906-27-7

IUPAC Name: N'-[(5-methylfuran-2-yl)methylidene]-2-phenoxybenzohydrazide
Synonyms: N'-[(5-methylfuran-2-yl)methylidene]-2-phenoxybenzohydrazide
Molecular Weight (g/mol): 320.35
Molecular Formula: C19H16N2O3
InChi Key: JJEDWBQZCRESJL-UHFFFAOYSA-N
SMILES: CC1=CC=C(O1)C=NNC(=O)C1=CC=CC=C1OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)